WPB01521WA
Whirl-Pak® Dosimeter Bag
W × L 15 cm × 25 cm , 6" x 10"
All Photos(1)
About This Item
Recommended Products
material
polyethylene
sterility
non-sterile
packaging
pkg of 500 ea
manufacturer/tradename
B01521WA (Nasco)
parameter
-346-180 °F temp. range (-210 to 82 °C)
W × L
15 cm × 25 cm , 6" x 10"
Related Categories
General description
These bags are specially designed to protect a dosimeter from moisture and contamination when working in a nuclear power plant or other hazardous area. The bold, red stripes make the bag highly visible so it will be seen quickly if it falls into the cooling water. The dosimeter can be read easily through the clear polyethylene, so there is no need to remove it. The extra long tabs on the bag make it convenient to attach to coveralls and other protective clothing. Fold or whirl the bag over to close and then twist the ends of the tab together to secure it in place. The strong polyethylene won′t break and is SEG incinerable. These smaller size bags are designed to hold the new, smaller dosimeters.
Legal Information
Whirl-pak is a registered trademark of The Aristotle Corporation