Key Documents
TMO362002-0008
Nalgene® Boston round bottle without cap
natural high-density polyethylene, volume 250 mL, case of 250 ea
Synonym(s):
HDPE bottle, nalgene bottle
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
Pricing and availability is not currently available.
material
natural high-density polyethylene
packaging
case of 250 ea
manufacturer/tradename
Nalgene 362002-0008
capacity
250 mL
volume
250 mL
General description
Conduct demanding packaging applications with Thermo Scientific Nalgene Boston Round HDPE Bottles without Closure, which are both durable and reliable products. These high density polyethylene bottles are rigid and translucent with excellent chemical resistance. Uniform walls make these bottles resistant to splitting and puncturing.
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Choose from one of the most recent versions:
Certificates of Analysis (COA)