All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C24H62O6Si6
CAS Number:
Molecular Weight:
615.26
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
storage temp.
2-8°C
SMILES string
C[Si](C)(C)OCC(O[Si](C)(C)C)C(O[Si](C)(C)C)C(O[Si](C)(C)C)C(CO[Si](C)(C)C)O[Si](C)(C)C
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service