Documents
T0443
Pharmed L/S tubing
size 13
Synonym(s):
Saint Gobain AY242606, PeristalticPump Tubing
About This Item
UNSPSC Code:
41105108
NACRES:
NB.35
packaging
pack of 25 feet
manufacturer/tradename
Tygon
I.D.
0.03 in. (0.8 mm)
General description
Tubing for MasterFlex Peristaltic Pumps, PharMed tubing has been specifically formulated for the MasterFlex pump system. Great for tissue and cell work- nontoxic and nonhemolytic. Long service life minimizes risk of fluid exposure; reduces tubing cost and pump downtime.
  • Opaque to UV and visible light to protect light-sensitive fluids
  • Heat sealable, bondable, and formable
  • Extremely low gas permeability
Legal Information
MasterFlex is a trademark of Cole-Parmer Instrument Co.