Skip to Content
Merck
All Photos(1)

Documents

SRP2117

Sigma-Aldrich

RNA Polymerase II, p14.5 subunit, GST tagged human

recombinant, expressed in E. coli, ≥85% (SDS-PAGE)

Synonym(s):

RPB9, hRPB14.5

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.26

biological source

human

recombinant

expressed in E. coli

Assay

≥85% (SDS-PAGE)

form

frozen liquid

mol wt

~42.2 kDa

packaging

pkg of 10 μg

storage condition

avoid repeated freeze/thaw cycles

concentration

250 μg/mL

color

clear colorless

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... POLR2I(5438)

Biochem/physiol Actions

hRPB9 is a subunit unique to RNA polymerase II, although it has sequence homologues in RNA polymerases I and III. The gene for Rpb9 is not essential for yeast cell viability, but is essential in Drosophila. hRPB9 has roles in both transcription initiation and transcription elongation. In the initiation reaction it is necessary for accurate start site selection. In the elongation reaction, RPB9, along with TFIIS facilitates the conversion of an arrest-competent conformation to a read-through competent conformation. RNA polymerase II lacking the RPB9 subunit uses alternate transcription initiation sites in vitro and in vivo and is unable to respond to the transcription elongation factor TFIIS in vitro. A role in the modulation of initiation and elongation is consistent with the localization of RPB9 in the three-dimensional structure of yeast RNA polymerase II. RPB9 is located at the tip of the so-called "jaws" of the enzyme, which is thought to function by clamping the DNA downstream of the active site. RPB9 comprises two zinc ribbon domains joined by a conserved linker region. The C-terminal zinc ribbon is similar in sequence to that found in TFIIS.

Physical form

Clear and colorless frozen liquid solution

Preparation Note

Use a manual defrost freezer and avoid repeated freeze-thaw cycles. While working, please keep sample on ice.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D A Harrison et al.
Molecular and cellular biology, 12(3), 928-935 (1992-03-01)
A small, divergently transcribed gene is located 500 bp upstream of the suppressor of Hairy-wing locus of Drosophila melanogaster. Sequencing of a full-length cDNA clone of the predominant 850-nucleotide transcript reveals that this gene encodes a 15,100-Da protein with high
Y Nogi et al.
Molecular and cellular biology, 13(1), 114-122 (1993-01-01)
We have previously isolated mutants of Saccharomyces cerevisiae that are primarily defective in transcription of 35S rRNA genes by RNA polymerase I and have identified genes (RRN1 to RRN9) involved in this process. We have now cloned the RRN4 gene
N A Woychik et al.
The Journal of biological chemistry, 266(28), 19053-19055 (1991-10-05)
The Saccharomyces cerevisiae RNA polymerase II subunit gene RPB9 was isolated and sequenced. RPB9 is a single copy gene on chromosome VII. The RPB9 sequence predicts a protein of 122 amino acids with a molecular mass of 14,200 Da. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service