Skip to Content
Merck
All Photos(1)

Documents

SRP0595

Sigma-Aldrich

GLS2, His-Tag

recombinant, expressed in E. coli

Synonym(s):

L-glutaminase, L-glutaminase amidohydrolase, glutaminase liver isoform (liver mitochondria)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.26

recombinant

expressed in E. coli

form

frozen liquid

mol wt

66 kDa

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... GLS2(27165)

General description

Human glutaminase liver isoform (liver, mitochondria), also known as GLS2, L-glutaminase, and L-glutaminase amidohydrolase, GenBank Accession No.NM_013267, a.a. 15-602, with N-terminal His-tag, MW=66 kDa, expressed in an E.coli cell expression system.

Physical form

Aqueous solution

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service