Skip to Content
Merck
All Photos(1)

Key Documents

SRP0211

Sigma-Aldrich

PTP-D2/PTPN14 Active human

recombinant, expressed in E. coli, ≥90% (SDS-PAGE)

Synonym(s):

Cytoskeletal-associated protein tyrosine phosphatase, PTP36, PTPN14, Pez, Protein tyrosine phosphatase, non-receptor type 14

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32
Pricing and availability is not currently available.

biological source

human

recombinant

expressed in E. coli

Assay

≥90% (SDS-PAGE)

form

aqueous solution

mol wt

61 kDa

packaging

pkg of 20 μg

storage condition

avoid repeated freeze/thaw cycles

concentration

>0.02 mg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... PTPN14(5784)

General description

Catalytic domain, (amino acids 817-1188), (Gene bank accession number NM_005401) with N-terminal GST tag, MW = 61 kDa, expressed in an E. coli expression system.

Application

Useful for the study of enzyme kinetics, regulation, to dephosphorylate target substrates and for screening inhibitors.

Unit Definition

One unit will hydrolyze 1 nmol p-nitrophenyl phosphate per minute at pH 7.4 and 30°C.

Physical form

Formulated in 25 mM Tris-HCl, pH 8.0, 75 mM NaCl, 0.05% Tween-20, 50% glycerol, 2 mM EDTA, 1 mM DTT, and 10 mM glutathione.

Preparation Note

Thaw on ice. Upon first thaw, briefly spin tube containing enzyme to recover full content of the tube. Aliquot enzyme into single use aliquots. Store remaining undiluted enzyme in aliquots at -70°C. Note: Enzyme is very sensitive to freeze/thaw cycles.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service