Skip to Content
Merck
All Photos(1)

Key Documents

SML1548

Sigma-Aldrich

THPN

≥98% (HPLC)

Synonym(s):

1-(2,4,6-Trihydroxyphenyl)-1-nonanone, 1-(3,4,5-Trihydroxyphenyl)nonan-1-one, CID 3063200

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H22O4
CAS Number:
Molecular Weight:
266.33
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 25 mg/mL, clear

storage temp.

2-8°C

SMILES string

OC1=C(O)C(O)=CC(C(CCCCCCCC)=O)=C1

InChI

1S/C15H22O4/c1-2-3-4-5-6-7-8-12(16)11-9-13(17)15(19)14(18)10-11/h9-10,17-19H,2-8H2,1H3

InChI key

NVFRHTFJDGAFQS-UHFFFAOYSA-N

General description

THPN or 1-(3,4,5-Trihydroxyphenyl)nonan-1-one, is a derivative of cytosporone B.

Biochem/physiol Actions

THPN is potent nuclear receptor TR3 (Nur77) antagonist that induces autophagic cell death in melanoma. Co-application of THPN with Akt2 inhibitors significantly represses tumor growth in xenograft mouse model.

Other Notes

air sensitive

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Impeding the interaction between Nur77 and p38 reduces LPS-induced inflammation.
Li L et al
Nature Chemical Biology, 11(5), 339-346 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service