Skip to Content
Merck
All Photos(1)

Documents

SML1478

Sigma-Aldrich

Ophiobolin A

≥95% (HPLC)

Synonym(s):

(2′S,3′S,3aR,5′R,6aS,9R,9aS,10aR)-1,3a,4,4′,5′,6a,7,8,9,9a,10,10a-Dodecahydro-9-hydroxy-3′,9,10a-trimethyl-5′-spiro[dicyclopenta[a,d]cyclooctene-3(2H),2′(3′H)-furan]-6-carboxaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H36O4
CAS Number:
Molecular Weight:
400.55
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

biological source

(from Biopolaris leersia fungal metabolite)

Quality Level

Assay

≥95% (HPLC)

form

powder

storage condition

protect from light

color

white to beige

storage temp.

−20°C

SMILES string

[H]C(/C1=C/C[C@]([C@]2(O[C@@H](C=C(C)C)C[C@@H]2C)CC3)([H])[C@@]3(C)C[C@@]([C@](C)(O)C4)([H])[C@]1([H])C4=O)=O

InChI

1S/C25H36O4/c1-15(2)10-18-11-16(3)25(29-18)9-8-23(4)12-19-22(20(27)13-24(19,5)28)17(14-26)6-7-21(23)25/h6,10,14,16,18-19,21-22,28H,7-9,11-13H2,1-5H3/b17-6-/t16-,18-,19-,21+,22+,23+,24+,25-/m0/s1

InChI key

MWYYLZRWWNBROW-BDZRSQQBSA-N

Biochem/physiol Actions

Ophiobolin A is a fungal metabolite, toxic to many crops and found to have antimicrobial and anticancer activity. Ophiobolin A is a calmodulin antagonist and inhibitor of big conductance Ca2+-activated K+ channel (BKCa) activity. Its anticancer activity may involve inhibition of multiple oncogenic signaling pathways including PI3K/mTOR, Ras/Raf/ERK and CDK/RB. One study suggested the possibility of pyrrolylation of lysine residues on its intracellular target protein(s) as a mechanism of action on various targets.

Other Notes

Light and air sensitive.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service