Skip to Content
Merck
All Photos(1)

Documents

SMB01057

Sigma-Aldrich

Pallidol

≥90% (LC/MS-ELSD)

Synonym(s):

Pallidol A

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H22O6
CAS Number:
Molecular Weight:
454.47
MDL number:
UNSPSC Code:
12352205
NACRES:
NA.25

biological source

plant

Assay

≥90% (LC/MS-ELSD)

form

solid

mol wt

454.47

solubility

water: slightly soluble

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

InChI

1S/C28H22O6/c29-15-5-1-13(2-6-15)23-25-19(9-17(31)11-21(25)33)28-24(14-3-7-16(30)8-4-14)26-20(27(23)28)10-18(32)12-22(26)34/h1-12,23-24,27-34H

InChI key

YNVJOQCPHWKWSO-UHFFFAOYSA-N

General description

Pallidol, a tetracyclic stilbenoid and a polyphenol, is a bioactive natural compound commonly derived from plants such as Cissus pallida, Chaenomeles sinensis, Gnetum latifolium, Pseudoalteromonas luteoviolacea, Caragana sinica, and Cissus repanda. Current research suggests that this metabolite acts as an inhibitor and may exhibit diverse biological activities, including anticancer, antioxidant, and antifungal properties.

Application

Pallidol is a natural product derived from plant source that finds application in compound screening libraries, metabolomics, phytochemical, and pharmaceutical research.

Features and Benefits

  • Suitable for Biochemical and Biomedical research
  • Versatile and adaptable for wide variety of laboratory and research applications

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service