RA481750-EP
Rotor for Eppendorf® 5810/5810R Centrifuges
A-4-81, includes 7x50 mL conical buckets
All Photos(1)
About This Item
Recommended Products
feature
to hold 0.2 to 500 mL tubes and bottles
A-4-81, includes 7x50 mL conical buckets
packaging
pack of 1 ea
pkg of 1 ea
manufacturer/tradename
Eppendorf® 022638607
parameter
4,000 max. RCF (3,220 xg)
technique(s)
centrifugation: suitable
compatibility
for use with centrifuge models 5810/5810R
Related Categories
General description
Rotor for Eppendorf(R) 5418/5418R Centrifuge
Legal Information
Eppendorf is a registered trademark of Eppendorf AG