Skip to Content
Merck
All Photos(1)

Documents

MSST0042

Sigma-Aldrich

SILuLite HAVCR1 Hepatitis A virus cellular receptor 1 human

recombinant, expressed in HEK 293 cells, MS Protein Standard

Synonym(s):

Kidney injury molecule 1 (KIM-1), T-cell immunoglobulin and mucin domain-containing protein 1 (TIMD-1), T-cell immunoglobulin mucin receptor 1 (TIM-1)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23201100
NACRES:
NA.12

biological source

human

Quality Level

recombinant

expressed in HEK 293 cells

tag

8-His tagged

Assay

≥98% (SDS-PAGE)

form

lyophilized powder

potency

≥98% Heavy amino acids incorporation efficiency by MS

mol wt

calculated mol wt 31 kDa

technique(s)

mass spectrometry (MS): suitable

suitability

suitable for mass spectrometry (internal calibrator)

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... HAVCR1(26762)

Related Categories

General description

HAVCR1 is a trans-membrane glycoprotein that is highly expressed in kidney proximal tubular cells with kidney injury. The KIM-1 ectodomain is cleaved, released into urine, and canbe quantified as a sensitive and specific biomarker for Acute Kidney Injury (AKI) in rodents and humans. In a recent study it was demonstrated that HAVCR1 is released into the circulation where it reports kidney injury in mice, rats, and humans with AKI and chronic kidney disease (CKD). Urinary HAVCR1 and MCP-1 levels, either alone or in combination, were significantly higher than those in AKI negative patients. Importantly, the combination of HAVCR1 and MCP-1 outperformed either biomarker alone.

Immunogen

SVKVGGEAGPSVTLPCHYSGAVTSMCWNRGSCSLFTCQNGIVWTNGTHVTYRKDTRYKLLGDLSRRDVSLTIENTAVSDSGVYCCRVEHRGWFNDMKITVSLEIVPPKVTTTPIVTTVPTVTTVRTSTTVPTTTTVPTTTVPTTMSIPTTTTVLTTMTVSTTTSVPTTTSIPTTTSVPVTTTVSTFVPPMPLPRQNHEPVATSPSSPQPAETHPTTLQGAIRREPTSSPLYSYTTDGNDTVTESSDGLWNNNQTQLFLEHSLLTANTTKGDYKDDDDKGHHHHHHHHGGQ

Biochem/physiol Actions

SILuLite HAVCR1 is a recombinant human protein expressed in human 293 cells. It is a protein consisting of 290 amino acids acids (the extracellular domain: Ser21-Gly290, and including C-terminal polyhistidine and FLAG® tags), with a calculated molecular mass of 31 kDa. SILu Lite HAVCR1 is an analytical standard designed to be used as starting material for preparation of calibrators and controls in LC-MS applications.

Physical form

Supplied as a lyophilized powder containing phosphate buffered saline.

Legal Information

FLAG is a registered trademark of Merck KGaA, Darmstadt, Germany
SILu is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service