Skip to Content
Merck
All Photos(1)

Documents

GE28-4039-43

Collection Plate

Cytiva 28-4039-43, pack of 1 ea

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
NACRES:
NA.56

material

polypropylene

feature

wetted part: no

packaging

pack of 1 ea

manufacturer/tradename

Cytiva 28-4039-43

Related Categories

General description

Collection Plate, 500 μL V-bottom.

Application

Collection plates is a package of 5 × 96-well plates designed for collection of eluted samples in parallel from screening plates such as MultiTrap and PreDictor plates. The volume in each well is 500 μL and the material in the collection plates is biocompatible polypropylene.

Features and Benefits

  • Convenient collection of samples in 96-well format
  • Excellent fit to prepacked MultiTrap plates and PreDictor plates
  • V-bottom for minimal loss of sample
  • Collection of < 500 μL volumes of sample per well

Legal Information

MultiTrap is a trademark of Cytiva

Storage Class Code

13 - Non Combustible Solids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service