Skip to Content
Merck
All Photos(1)

Documents

GE17549853

HiTrap® MabSelect PrismA

Cytiva, 17549853, pack of 1 × 5 mL

Synonym(s):

MabSelect PrismA

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106500
NACRES:
NB.21

packaging

pack of 1 × 5 mL

manufacturer/tradename

17549853
Cytiva

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

These HiTrap columns are prepacked with MabSelect PrismA protein A chromatography resin. This affinity resin in the packed column has been improved with an optimized high-flow agarose base matrix and a genetically engineered protein ligand, allowing efficient cleaning between monoclonal antibody purification runs. This allows future demands in monoclonal antibody purification to be met, including processing of many bispecific antibodies.

The enhanced alkaline stability of MabSelect PrismA also provides opportunities for more robust mAb purification using periodic counter-current chromatography. The possibility for cleaning using 1 M NaOH increases bioburden control for continuous processes, which can be exposed to days or weeks of feed harvest.

Features and Benefits

  • Enhanced binding capacity compared with other Protein A resins
  • Excellent alkaline stability enables efficient cleaning and sanitization using 0.5–1.0 M NaOH
  • Covered by a comprehensive security of supply program, including dual sources of the agarose base matrix and protein A ligand.
  • The PrismA protein A ligand is also available in HiTrap® Fibro and HiScreen Fibro PrismA chromatography units for ultrafast purification of mAbs.
  • Easily measure protein A leakage using the PrismA ELISA kit specifically designed for use with MabSelect PrismA resins.

Legal Information

HiScreen is a trademark of Cytiva
HiTrap is a registered trademark of Cytiva
MabSelect is a trademark of Cytiva

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service