Skip to Content
Merck
All Photos(1)

Documents

GE17-0485-03

Cytodex® 3 microcarrier beads

Cytiva, pkg of 500 g

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41121800
NACRES:
NC.01

material

dextran beads

sterility

non-sterile

packaging

pkg of 500 g

manufacturer/tradename

17-0485-03
Cytiva

Looking for similar products? Visit Product Comparison Guide

General description

The microcarrier of choice for cells that may be difficult to culture in vitro.

Microcarrier culture is a technique which makes possible the practical high yield culture of anchorage-dependent cells. Cytodex has been specifically developed for the culture of a wide range of animal cells in culture volumes ranging from a few ml to more than 6000 L. Using Cytodex in simple suspension culture systems provides yields of several million cells per mL.

Formed by chemically coupling a thin layer of denatured collagen to the cross-linked dextran matrix, this is the microcarrier of choice for cells that may be difficult to culture in vitro, and particularly for cells with an epithelial like morphology.

Application

Cytodex™3 microcarrier beads are suitable for:
  • the isolation, transportation, and storage of cells
  • cells that might be challenging to culture in vitro, especially cells with an epithelial morphology, sensitive, differentiated, or low in plating efficiency (e.g., endocrine cells, hepatocytes)
  • large-scale production of viruses, cells, and recombinant cell products (e.g.  hormones, nucleic acids, enzymes, interferon)

Features and Benefits

  • Designed for culture of primary cells or those with an epithelioid morphology, especially cells with low plating-efficiency or differentiated or sensitive cell-types which are normally difficult to grow, e.g. hepatocytes, endocrine cells.
  • Contains a layer of acid-denatured porcine collagen (gelatin) on the surface of the microporous matrix.
  • Designed to yield harvested cells of the highest viability.
  • Transparent for easy microscopic examination of attached cells.

Preparation Note

Prior to use, swell beads in PBS (50 mL/g) and autoclave at 120°C.

Legal Information

Cytodex is a registered trademark of Cytiva

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Atorvastatin and conditioned media from atorvastatin-treated human hematopoietic stem/progenitor-derived cells show proangiogenic activity in vitro but not in vivo
Taha and Witold N ,Nowak
Mediators of Inflammation, 2019-2019 (2019)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service