Skip to Content
Merck
All Photos(1)

Documents

G2549

SAFC

Glasgow Minimum Essential Medium

With sodium bicarbonate and ʟ-glutamine, sterile liquid, suitable for cell culture

Synonym(s):

GMEM, Glasgow′s MEM

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352207
NACRES:
NA.75

product name

Glasgow Minimum Essential Medium, With sodium bicarbonate and L-glutamine, liquid, sterile-filtered, suitable for cell culture

Quality Level

sterility

sterile-filtered

form

liquid

technique(s)

cell culture | mammalian: suitable

impurities

Endotoxin, tested

components

phenol red: 0.0213 g/L
L-glutamine: 0.292 g/L
NaHCO3: 2.75 g/L
glucose: 4.5 g/L (Dextro)

shipped in

ambient

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

For use with adherent kidney cell lines such as baby hamster kidney cells (BHK).

Preparation Note

Supplement with Tryptose phosphate broth solution (T8159) at 100 ml/L of medium.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Augmentation of biological titer of foot and mouth disease virus in in vitro culture.
Akram Q
The Journal of Animal and plant sciences, 23, 771-774 (2013)
Edward C Hsiao et al.
PloS one, 3(7), e2532-e2532 (2008-07-04)
Fluorescent reporters are useful for assaying gene expression in living cells and for identifying and isolating pure cell populations from heterogeneous cultures, including embryonic stem (ES) cells. Multiple fluorophores and genetic selection markers exist; however, a system for creating reporter
Linda Cambier et al.
Development (Cambridge, England), 141(15), 2959-2971 (2014-07-24)
A complex regulatory network of morphogens and transcription factors is essential for normal cardiac development. Nkx2-5 is among the earliest known markers of cardiac mesoderm that is central to the regulatory pathways mediating second heart field (SHF) development. Here, we

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service