EP5910705000
Adapter for Eppendorf® S-4x400 Rotor
for 250 mL bottle, pkg of 2 ea
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
feature
for 250 mL bottle
packaging
pkg of 2 ea
manufacturer/tradename
Eppendorf® 5910705000
technique(s)
centrifugation: suitable
compatibility
for use with 400 mL round bucket
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Adapter for various size tubes and bottles for use with Eppendorf S-4x400 rotor.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service