Skip to Content
Merck
All Photos(1)

Documents

EP0030122704

Eppendorf® Screw Caps for 25 & 50 mL Conical Tubes

PCR clean, non-sterile, pack of 10 × 50 ea

Synonym(s):

Screw Caps for tubes, Screw covers

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.11

sterility

non-sterile

feature

DNase free
PCR clean

packaging

pack of 10 × 50 ea

manufacturer/tradename

Eppendorf® 0030122704

suitability

suitable for PCR

Application

PCR

Legal Information

Eppendorf is a registered trademark of Eppendorf AG

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service