Skip to Content
Merck

C2513

Sigma-Aldrich

(±)-Citronellal

≥85%

Synonym(s):

(±)-3,7-Dimethyl-6-octenal

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C=CHCH2CH2CH(CH3)CH2CHO
CAS Number:
Molecular Weight:
154.25
Beilstein:
1720789
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Pricing and availability is not currently available.

Assay

≥85%

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.451 (lit.)

bp

207 °C (lit.)

density

0.857 g/mL at 25 °C (lit.)

application(s)

forensics and toxicology
veterinary

SMILES string

CC(CC\C=C(\C)C)CC=O

InChI

1S/C10H18O/c1-9(2)5-4-6-10(3)7-8-11/h5,8,10H,4,6-7H2,1-3H3

InChI key

NEHNMFOYXAPHSD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
WH0054708M6SAB2101432HPA043406
antibody form

affinity isolated antibody

antibody form

purified immunoglobulin

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

species reactivity

mouse, rat, bovine, dog, rabbit, horse, guinea pig, human

species reactivity

human

species reactivity

rat, human, guinea pig, horse, rabbit, dog, mouse, bovine

species reactivity

human

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

Gene Information

human ... MARCH5(54708)

Gene Information

human ... MARCH5(54708)

Gene Information

human ... MARCH1(55016)

Gene Information

human ... MARCH3(115123)

technique(s)

western blot: suitable

technique(s)

indirect ELISA: suitable

technique(s)

western blot: suitable

technique(s)

immunoblotting: 0.04-0.4 μg/mL, immunofluorescence: 0.25-2 μg/mL, immunohistochemistry: 1:50-1:200

replaced by

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1B

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

165.2 °F - closed cup

Flash Point(C)

74 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Need A Sample COA?

This is a sample Certificate of Analysis (COA) and may not represent a recently manufactured lot of this specific product.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Martin F Strube-Bloss et al.
PloS one, 10(9), e0137413-e0137413 (2015-09-05)
To trigger innate behavior, sensory neural networks are pre-tuned to extract biologically relevant stimuli. Many male-female or insect-plant interactions depend on this phenomenon. Especially communication among individuals within social groups depends on innate behaviors. One example is the efficient recruitment

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service