Skip to Content
Merck
All Photos(1)

Documents

A5711

Sigma-Aldrich

Arachidonic acid-5,6,8,9,11,12,14,15-3H(N)

≥95% (Radiochemical Purity), ethanol solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H32O2
CAS Number:
Molecular Weight:
304.47
MDL number:
UNSPSC Code:
12352200

Assay

≥95% (Radiochemical Purity)

form

ethanol solution

extent of labeling

180-240 Ci/mmol

mass shift

M+16

storage temp.

−20°C

SMILES string

[3H]\C(CCCCC)=C(/[3H])C\C([3H])=C(\[3H])C\C([3H])=C(\[3H])C\C([3H])=C(\[3H])CCCC(O)=O

Looking for similar products? Visit Product Comparison Guide

Packaging

serum bottle

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service