Skip to Content
Merck
All Photos(1)

Key Documents

A1433

Sigma-Aldrich

Ammonium persulfate

ACS reagent, ≥98.0%

Synonym(s):

AP, APS, Ammonium peroxodisulfate, Ammonium peroxydisulfate, PER

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(NH4)2S2O8
CAS Number:
Molecular Weight:
228.20
EC Number:
MDL number:
UNSPSC Code:
12352300

grade

ACS reagent

vapor density

7.9 (vs air)

Assay

≥98.0%

impurities

≤0.005% Insoluble matter
≤0.04 meq/g Titratable free acid

ign. residue

≤0.05%

anion traces

chloride, chlorate (as Cl-): ≤0.001%

cation traces

Fe: ≤0.001%
Mn: ≤0.5 ppm
heavy metals (as Pb): ≤0.005%

SMILES string

N.N.OS(=O)(=O)OOS(O)(=O)=O

InChI

1S/2H3N.H2O8S2/c;;1-9(2,3)7-8-10(4,5)6/h2*1H3;(H,1,2,3)(H,4,5,6)

InChI key

ROOXNKNUYICQNP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Catalyst for acrylamide gel polymerization.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Ox. Sol. 3 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service