Skip to Content
Merck
All Photos(1)

Key Documents

Z801585

Hellma® calibration standard, glass filter set

F3, F4, F7, for testing the wavelength accuracy and the photometric accuracy

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12000000
NACRES:
SB.32

grade

certified reference material

manufacturer/tradename

Hellma (666-S001)

application(s)

food and beverages
petroleum
pharmaceutical
plastics

format

neat

Looking for similar products? Visit Product Comparison Guide

General description

Secondary calibration standard for wavelength accuracy and photometric accuracy. Certified.
Glass Filter Set: F3, F4, F7 (peak positions in nm, A: 270; 280; 297; 320; 340; 440; 465; 546,1; 590; 635; W: 329; 472; 512; 681; 875)

Other Notes

The most current version of Hellma's handling guidelines for calibration standards can be accessed on the Hellma Analytics web site.

Legal Information

Hellma is a registered trademark of Hellma GmbH

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service