SIAL113AA500
Amber Wide-Mouth Boston Round Bottle, HDPE
capacity 500 mL
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121800
material
amber bottle
amber cap
high-density polyethylene bottle
polypropylene cap
round bottle
wide-mouth bottle
packaging
pkg of 4 × 12 ea
manufacturer/tradename
Sigma-Aldrich
H
170 mm
capacity
500 mL
diam.
72.7 mm
fitting
53-415
Looking for similar products? Visit Product Comparison Guide
General description
Protect light-sensitive materials during shipping or storage. Wide mouth is easy to fill. Opaque Amber reduces UV light transmissions to protect light-sensitive liquids. Excellent chemical resistance to most acids, bases and alcohols make these versatile bottles where exposure to light needs to be restricted.
Includes: Amber polypropylene screw caps. Linerless caps provide leakproof protection without the use of liner that can wrinkle, cause leaks, or contaiminate your reagents.
Includes: Amber polypropylene screw caps. Linerless caps provide leakproof protection without the use of liner that can wrinkle, cause leaks, or contaiminate your reagents.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service