All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C37H39N5O17
CAS Number:
Molecular Weight:
825.73
MDL number:
UNSPSC Code:
12171500
Assay
≥85% (HPLC)
storage temp.
−20°C
Application
Cell impermeable fluorescent probe for Ca2+; compared with other probes, there is less compartmentalization and leakage; cells remain brightly fluorescent and responsive to changes in cytosolic free Ca2+ for hours.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service