Skip to Content
Merck
All Photos(1)

Key Documents

CRM42173

Supelco

Hexachlorobenzene

certified reference material

Synonym(s):

HCB

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6Cl6
CAS Number:
Molecular Weight:
284.78
Beilstein:
1912585
MDL number:
UNSPSC Code:
41116107
Pricing and availability is not currently available.

grade

certified reference material
TraceCERT®

product line

TraceCERT®

bp

323-326 °C (lit.)

mp

227-229 °C (lit.)

storage temp.

2-8°C

SMILES string

Clc1c(Cl)c(Cl)c(Cl)c(Cl)c1Cl

InChI

1S/C6Cl6/c7-1-2(8)4(10)6(12)5(11)3(1)9

InChI key

CKAPSXZOOQJIBF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Footnote

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - STOT RE 1 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service