Skip to Content
Merck
All Photos(1)

Key Documents

A2931

Sigma-Aldrich

Alkali Blue 6B

Synonym(s):

Acid Blue 110, Acid Blue 119

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C37H31N3O4S
CAS Number:
Molecular Weight:
613.72
Colour Index Number:
42750
EC Number:
UNSPSC Code:
12171500

composition

Dye content, ≥80%

InChI

1S/C37H29N3O3S/c41-44(42,43)36-14-8-7-13-35(36)40-34-25-19-29(20-26-34)37(27-15-21-32(22-16-27)38-30-9-3-1-4-10-30)28-17-23-33(24-18-28)39-31-11-5-2-6-12-31/h1-26,38-39H,(H,41,42,43)

InChI key

DVPLSZDCYOJSOG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service