Skip to Content
Merck
All Photos(1)

Documents

70200

Supelco

AQUANAL-professional Vario H Chlorine Total (DPD)

determination of chlorine

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41113319

form

powder

measuring range

0.01-2 mg/L (Cl2)

application(s)

environmental
general analytical

Packaging

100 powder packs for 10 mL sample volume

Analysis Note

method: N,N-Diethyl-p-phenylenediamine (DPD)

Legal Information

AQUANAL is a trademark of Sigma-Aldrich International GmbH

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Klaudia Bednarz et al.
Frontiers in neural circuits, 14, 21-21 (2020-06-12)
Neuroendocrine circuits are orchestrated by the pituitary gland in response to hypothalamic hormone-releasing and inhibiting factors to generate an ultradian and/or circadian rhythm of hormone secretion. However, mechanisms that govern this rhythmicity are not fully understood. It has been shown

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service