Skip to Content
Merck
All Photos(1)

Key Documents

69894

Sigma-Aldrich

MES potassium salt

≥99.0% (T)

Synonym(s):

2-(N-Morpholino)ethanesulfonic acid potassium salt, 4-Morpholineethanesulfonic acid potassium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12KNO4S
CAS Number:
Molecular Weight:
233.33
MDL number:
UNSPSC Code:
12161700
Pricing and availability is not currently available.

Assay

≥99.0% (T)

impurities

≤1% water

useful pH range

5.5-6.7

pKa 

6.1

solubility

H2O: 0.35 g/mL, clear, colorless

SMILES string

[K+].[O-]S(=O)(=O)CCN1CCOCC1

InChI

1S/C6H13NO4S.K/c8-12(9,10)6-3-7-1-4-11-5-2-7;/h1-6H2,(H,8,9,10);/q;+1/p-1

InChI key

IMFIKFZWLAWMQE-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service