Skip to Content
Merck
All Photos(2)

Key Documents

55591

Supelco

Oxymetazoline hydrochloride

analytical standard

Synonym(s):

2-(3-Hydroxy-2,6-dimethyl-4-t-butylbenzyl)-2-imidazoline hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H24N2O · HCl
CAS Number:
Molecular Weight:
296.84
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

Assay

≥99.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤0.5% water

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

SMILES string

Cl.Cc1cc(c(O)c(C)c1CC2=NCCN2)C(C)(C)C

InChI

1S/C16H24N2O.ClH/c1-10-8-13(16(3,4)5)15(19)11(2)12(10)9-14-17-6-7-18-14;/h8,19H,6-7,9H2,1-5H3,(H,17,18);1H

InChI key

BEEDODBODQVSIM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Oxymetazoline hydrochloride may be used as a reference standard in the determination of oxymetazoline hydrochloride in nasal spray formulations using high performance liquid chromatography (HPLC).[1]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

Oxymetazoline hydrochloride is a partial α2A-adrenoceptor agonist; agonist at 5-HT1A, 5-HT1B, and 5-HT1D serotonin receptors. It is a mixed agonist-antagonist at 5-HT1C serotonin receptors.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 1 Oral - Aquatic Chronic 3 - Eye Dam. 1

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simultaneous determination of triamcinolone acetonide and oxymetazoline hydrochloride in nasal spray formulations by HPLC.
Sudsakorn S, et al.
Journal of Pharmaceutical and Biomedical Analysis, 40(5), 1273-1280 (2006)
Sylwia K Paszun et al.
Acta poloniae pharmaceutica, 70(6), 1079-1085 (2014-01-05)
The presented study aimed at the evaluation of hydrochlorothiazide influence on cilazapril stability in model mixture and fixed dose tablet formulation. The degradation of cilazapril in the presence of hydrochlorothiazide took place according to autocatalytic reaction kinetic mechanism, described mathematically

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service