Skip to Content
Merck
All Photos(1)

Documents

31666

Supelco

Prosulfuron

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H16F3N5O4S
CAS Number:
Molecular Weight:
419.38
Beilstein:
7800481
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

COc1nc(C)nc(NC(=O)NS(=O)(=O)c2ccccc2CCC(F)(F)F)n1

InChI

1S/C15H16F3N5O4S/c1-9-19-12(22-14(20-9)27-2)21-13(24)23-28(25,26)11-6-4-3-5-10(11)7-8-15(16,17)18/h3-6H,7-8H2,1-2H3,(H2,19,20,21,22,23,24)

InChI key

LTUNNEGNEKBSEH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Prosulfuron is a selective, sulfonylurea herbicide, used in controlling broadleaf weeds in agricultural crops.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Seunghun Hyun et al.
Journal of environmental quality, 33(4), 1354-1361 (2004-07-16)
Prosulfuron [1-(4-methoxy-6-methyltriazin-2-yl)-3-[2-(3,3,3-trifluoropropyl) phenylsulfonyl]-urea), a relatively new sulfonylurea herbicide, is a weak acid (pK(a) 3.76), and therefore, will undergo pH-dependent speciation and sorption. Understanding prosulfuron sorption in soils is important for predicting its environmental fate. Soil and solution factors controlling sorption
Ryan P Hultgren et al.
Journal of agricultural and food chemistry, 50(11), 3236-3243 (2002-05-16)
The sulfonylurea herbicide prosulfuron, 1-(4-methoxy-6-methyltriazin-2-yl)-3-[2-(3,3,3-trifluoropropyl)phenylsulfonyl]urea, is used for the selective control of broadleaf weeds in corn, sorghum, and cereal grains. To investigate its fate in soils, this study examined the effects of soil pH and water content on the rates
Seunghun Hyun et al.
Environmental science & technology, 38(20), 5413-5419 (2004-11-17)
Sorption of organic acids by variable-charge soil occurs through both hydrophilic and hydrophobic sorption. In this study, the effect of chemical acidity and the type of acidic functional group on the relative contribution of hydrophilic and hydrophobic processes to sorption
Christophe Menniti et al.
Journal of agricultural and food chemistry, 51(12), 3525-3527 (2003-05-29)
The transformation of prosulfuron [1-(4-methoxy-6-methyltriazine-2-yl)-3-[2-(3,3,3-trifluropropyl)phenylsulfonyl]urea] in three soils at different pH values (sterilized and unsterilized) was studied, and it was shown that the rate of transformation was high in acidic soil. From the results obtained in sterile soils, it is
Hydrolysis of prosulfuron at pH 5: evidence for a resonance-stabilized triazine cleavage product
Bray.DL, et al.
Pestic. Sci., 51(1), 56-64 (1997)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service