Skip to Content
Merck
All Photos(1)

Documents

31476

Supelco

Benzoylprop-ethyl

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H17Cl2NO3
CAS Number:
Molecular Weight:
366.24
Beilstein:
2899940
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CCOC(=O)C(C)N(C(=O)c1ccccc1)c2ccc(Cl)c(Cl)c2

InChI

1S/C18H17Cl2NO3/c1-3-24-18(23)12(2)21(14-9-10-15(19)16(20)11-14)17(22)13-7-5-4-6-8-13/h4-12H,3H2,1-2H3

InChI key

SLCGUGMPSUYJAY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Benzoylprop-ethyl may be used as a reference standard for the determination of the analyte:
  • In various fruits and vegetables by gas chromatography/mass spectrometry (GC/MS) and liquid chromatography with fluorescence detection (LC-FLD).
  • In animal tissues by gel permeation chromatography (GPC) cleanup followed by GC–MS and liquid chromatography-tandem mass spectrometry (LC–MS–MS).

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Multiresidue method for the determination of residues of 251 pesticides in fruits and vegetables by gas chromatography/mass spectrometry and liquid chromatography with fluorescence detection.
Fillion J, et al.
Journal of AOAC (Association of Official Analytical Chemists) International, 83(3), 698- 713 (2000)
Comparison of liquid and gas chromatography for the determination of bromoxynil octanoate and benzoylprop ethyl in wheat products.
J F Lawrence et al.
Journal of agricultural and food chemistry, 28(5), 1019-1022 (1980-09-01)
Validation study on 660 pesticide residues in animal tissues by gel permeation chromatography cleanup/gas chromatography-mass spectrometry and liquid chromatography-tandem mass spectrometry.
Pang GF, et al.
Journal of Chromatography A, 1125(1), 1-30 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service