Skip to Content
Merck
All Photos(1)

Documents

07391

Supelco

Glycolate Standard for IC

TraceCERT®, 1000 mg/L glycolate in water (nominal concentration)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

concentration

1000 mg/L glycolate in water (nominal concentration)

technique(s)

ion chromatography: suitable

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

format

single component solution

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to SI unit kg and measured against primary material from an NMI, e.g. NIST.
Certified content incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com.

Linkage

Visit the IC Portal to learn more

Analysis Note

1000 mg/L glycolate in water, prepared with glycolic acid and water; filtered through a 0.45 μm membrane filter and stabilized with sodium azide

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service