Skip to Content
Merck
All Photos(1)

Key Documents

28-3320

Sigma-Aldrich

Sodium metaborate tetrahydrate

CP

Synonym(s):

Boric acid, sodium salt, tetrahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaBO2 · 4H2O
CAS Number:
Molecular Weight:
137.86
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

grade

CP

form

solid

availability

available only in Japan

SMILES string

O.O.O.O.[Na+].[O-]B=O

InChI

1S/BO2.Na.4H2O/c2-1-3;;;;;/h;;4*1H2/q-1;+1;;;;

InChI key

JAKYJVJWXKRTSJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service