Documents
ZFC0NDCL1
Milli-Q
Feed Water Conductivity Cell
for use with Milli-Q® Reference, To monitor the ionic quality of feed water
About This Item
UNSPSC Code:
41104200
NACRES:
JA.13
packaging
pkg of 1 unit
manufacturer/tradename
Milli-Q®
H × W × D
3.5 cm (1.4 in.) × 8.5 cm (3.3 in.) × 6.5 cm (2.5 in.)
diam.
23 cm (9 in.)
compatibility
for use with Milli-Q® Reference
General description
The feed water conductivity cell is an installation accessory which triggers an alarm if the feed water quality is out of specifications. This prevents premature exhaustion of the Q-Gard® and Quantum® cartridges.
Application
The feed water conductivity cell is used to monitor the feed water ionic quality for Milli-Q® Reference system.
Legal Information
Milli-Q is a registered trademark of Merck KGaA, Darmstadt, Germany
Q-GARD is a registered trademark of Merck KGaA, Darmstadt, Germany
QUANTUM is a registered trademark of Merck KGaA, Darmstadt, Germany