Skip to Content
Merck
All Photos(1)

Documents

SX1330

Supelco

Spill Kit, Solvent Spill Adsorber

SpillSolv®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
47131905

H × L × W

13 in. × 19½ in. × 7 in.

shipped in

ambient

storage temp.

room temp

Components

6 x 0.5 kg SpillSolv Volatile Adsorbent Powder, 1 pair splash goggles with ventilation, 1 pair nitril gloves, 6 H.D. disposal bags 14" x 20" x 4 mil, 4 disposable plastic scoops and scrapers, 1 big sponge for residue clean up and 1 treatment guide.

Legal Information

SPILLSOLV is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service