Skip to Content
Merck
All Photos(2)

Documents

MABS1255

Sigma-Aldrich

Anti-Platelet Factor 4 Antibody, clone 197.2

clone 197.2, from mouse

Synonym(s):

Platelet factor 4, C-X-C motif chemokine 4, Iroplact, Oncostatin-A, PF-4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

197.2, monoclonal

species reactivity

human

technique(s)

ELISA: suitable
flow cytometry: suitable
immunocytochemistry: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... PF4(5196)

Related Categories

General description

Platelet factor 4 (UniProt P02776; also known as C-X-C motif chemokine 4, Iroplact, Oncostatin-A, PF-4) is encoded by the PF4 (also known as CXCL4, SCYB4) gene (Gene ID 5196) in human. Platelet factor 4 (PF4) is a small CXC chemokine family cytokine released from alpha-granules of activated platelets during platelet aggregation to promote blood coagulation. PF4 binds heparin with high affinity (Kd ~4-20 nM) via its heparin-binding domain (a.a. 92-98) and neutralizes the anti-thrombin activity of heparin-like molecules. PF4 is chemotactic for neutrophils, fibroblasts and monocytes, and interacts with the chemokine receptor CXCR3 splice variant CXCR3B. In individuals suffering from heparin-induced thrombocytopenia (HIT), PF4-heparin complexes can trigger an immune response and the generation of anti-PF4 antibodies, resulting in the foramtion of macromolecular antibody-heparin-PF4 immune complexes that bind Fc RIIa (CD32a) on platelets and induce platelet activation. Platelet activation in turn results in more PF4 release into circulation, enhanced immune-complex formation, exaggerated platelet activation, and cell clearance. PF4 is synthesized with a signal peptide sequence (a.a. 1-31) that is removed posttranslationally to produce the mature PF4 (a.a. 32-101), additional proteolytic cleavage yields the PF4 short form (a.a. 48-101).

Specificity

Clone 197.2 targets a linear epitope sensitive to heparin binding, but insensitive to reducing agent or denaturation by SDS (Xiao, Z., et al. (2008). Blood. 112(4):1091-1100).

Immunogen

Purified human platelet PF4 (Xiao, Z., et al. (2008). Blood. 112(4):1091-1100).

Application

Detect PF4 using this mouse monoclonal Anti-Platelet Factor 4, clone 197.2, Cat. No. MABS1255, validated for use in ELISA, Flow Cytometry, Function Analysis, Immunocytochemistry. and Western Blotting.
Western Blotting Analysis: A representative lot detected E. coli expressed recombinant mature human PF4 (a.a. 32-101) under both reduced and non-reduced condition (Courtesy of Dr. Richard H. Aster, MD, PhD, Blood Center of Wisconsin, U. S. A.).

ELISA Analysis: A representative lot detected PF4 in the absence of heparin by ELISA. Decreased immunoreactivity of clone 197.2 was seen toward heparin-complexed PF4 (Xiao, Z., et al. (2008). Blood. 112(4):1091-1100).

Function Analysis: A representative lot and PF4, but not clone 197.2 or PF4 alone, activated human neutrophils as indicated by Mac-1 upregulation (Xiao, Z., et al. (2008). Blood. 112(4):1091-1100).

Immunocytochemistry Analysis: A representative lot, preconjugated with Alexa Fluor 488, immunolabeled the co-administered PF4 on human neutrophils. CD32a (Fc gamma RIIA) was seen co-localized with the PF4 anti-PF4 immune complexes on the surface of activated neutrophils (Xiao, Z., et al. (2008). Blood. 112(4):1091-1100).

Western Blotting Analysis: A representative lot detected PF4 under both reduced and non-reduced condition by recognizing a linear epitope (Xiao, Z., et al. (2008). Blood. 112(4):1091-1100).

Quality

Evaluated by Flow Cytometry in human platelets.

Flow Cytometry Analysis: 1 µg of this antibody detected PF4 in one million 4% paraformaldehyde-fixed, 0.2% Triton X-100-permeabilized human platelets.

Target description

~8 kDa (recombinant mature hPF4) observed. 10.84 kDa (pro-form), 7.769 kDa (mature; a.a. 32-101), 6.033 kDa (cleaved short form; a.a. 48-101) calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Purified mouse IgG1 in buffer containing PBS without preservatives.

Other Notes

Concentration: Please refer to lot specific datasheet.

Legal Information

ALEXA FLUOR is a trademark of Life Technologies

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service