Skip to Content
Merck
All Photos(3)

Documents

MABC28

Sigma-Aldrich

Anti-RIP3 Antibody, clone 6E6.2

clone 6E6.2, from mouse

Synonym(s):

Receptor-interacting serine/threonine-protein kinase 3, RIP-like protein kinase 3, Receptor-interacting protein 3, RIP-3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

6E6.2, monoclonal

species reactivity

human, rat

technique(s)

immunohistochemistry: suitable
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... RIPK3(11035)

General description

Receptor-interacting protein 3 (RIP3) contains a N-terminal kinase domain and interacts with RIP. The binding of RIP to RIP3 causes the RIP3/RIP complex to be shuttled into the tumor necrosis factor receptor-1 signaling cascade. RIP3 is known to be a very effective apoptosis inducer. Two novel splice variants of RIP3 have been discovered, RIP3 beta and RIP3 gamma, that have shown to decrease apoptosis related to full length RIP3. RIP3 gamma has been shown to be significantly up-regulated in colon and lung cancers and may play a role in tumorigenesis.

Specificity

This antibody recognizes RIP3 at the N-terminus.

Immunogen

Epitope: N-terminus
GST-tagged recombinant protein corresponding to human RIP3 at the N-terminus.

Application

Research Category
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional
This Anti-RIP3 Antibody, clone 6E6.2 is validated for use in WB, IH for the detection of RIP3.
Western Blot Analysis: A 1:1,000 dilution from a representative lot detected RIP3 in 10 µg of human placenta tissue lysate.

Immunohistochemistry Analysis: A 1:500 dilution from a representative lot detected RIP3 in rat pancreas tissue.

Quality

Evaluated by Western Blot in human pancreas tissue lysate.

Western Blot Analysis: A 1:1,000 dilution of this antibody detected RIP3 in 10 µg of human pancreas tissue lysate.

Target description

~56 kDa (isoform 1) and ~27 kDa (isoform 2) observed.
Isoform 3 (~25 kDa) may be observed in some lysates. An uncharacterized band may be observed at ~16 kDa in some lysates.

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG2aκ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Human pancreas tissue lysate

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anne-Sophie Gary et al.
Cell death discovery, 10(1), 232-232 (2024-05-15)
Ultraviolet radiation (UVR) is a major environmental mutagen. In skin, UVR can initiate cancer through the induction of mutagenic DNA damage and promote its progression. An important cancer prevention mechanism is the regulated cell death (RCD), which can safely dispose

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service