Skip to Content
Merck
All Photos(1)

Documents

ALP30

Sigma-Aldrich

Apolipoprotein B, human

Synonym(s):

ApoB

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.41

biological source

human

Quality Level

Assay

>95% (SDS-PAGE)

form

solid

manufacturer/tradename

Chemicon®

NCBI accession no.

UniProt accession no.

shipped in

dry ice

Gene Information

human ... APOB(338)

General description

Product Source: Human plasma, fresh, non-frozen. Tested negative for anti-HIV-1, anti-HIV-2, anti-HCV, anti-HBc and HBsAg.
Two forms of Apo B exist: Apo B-100 and Apo B-48. The first is found in VLDL and LDL and is produced by the liver. The second is found in chylomicrons and originates in the intestine.

Apo B is purified from LDL isolated by density gradient ultracentrifugation (1.03-1.05 g/mL density range) followed by size exclusion chromatography in 10 mM deoxycholate, 50 mM NaCl, 50 mM Na2CO3, pH 10 (Walsh & Atkinson 1983).

PROPERTIES:

Apo B, which has a Mr of 550 kD, is the major protein found in LDL and is responsible for the cellular uptake of LDL particles from the plasma (Goldstein & Brown 1982). The Apo B prepared by this procedure is aggregated. The aggregates are not retained by a 0.45 mm filter, but migrate only a short distance into a 5% SDS PAGE gel. In the isolation buffer, Apo B gives a stable solution that is slightly hazy.

Physical form

Lyophilized from 50 mM Na2CO3, pH 10, 50 mM NaCl, and 10 mM sodium deoxycholate solution. Non-sterile.

Storage and Stability

Maintain lyophilized material at <= -20°C for up to six months. Reconstitute to 1 mL by adding deionized water. If the deoxycholate is removed, Apo B-100 will slowly aggregate and become insoluble. After reconstitution maintain at +2-8°C for up to 4 weeks.

Analysis Note

Single arc by IEP against antisera to whole human serum.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service