Skip to Content
Merck
All Photos(1)

Key Documents

AB1923

Sigma-Aldrich

Anti-Integrin αV Antibody, cytoplasmic domain

serum, Chemicon®

Synonym(s):

Vitronectin Receptor alpha Subunit, CD51 Antigen

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

Gene Information

human ... ITGAV(3685)

Specificity

Integrin alphaV

Immunogen

Epitope: cytoplasmic domain
Synthetic peptide corresponding to amino acids 1035-1048 of human integrin alphaV. This peptide sequence is absolutely conserved in the mouse integrin alphaV sequence.

Application

Anti-Integrin αV Antibody, cytoplasmic domain detects level of Integrin αV & has been published & validated for use in ELISA & WB.
ELISA: 1:500 (direct)

Western Blot: 1:500. Antibody detects a non-reducible 155 kDa band corresponding to integrin alphaV.

Optimal dilutions must be determined by the end user.
Research Category
Cell Structure
Research Sub Category
Integrins

Physical form

Whole rabbit serum. Liquid containing no preservative

Storage and Stability

Maintain at -20°C in undiluted aliquots for up to 12 months from date of receipt. Avoid repeated freeze/thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Syndecan-1 regulates alphavbeta3 and alphavbeta5 integrin activation during angiogenesis and is blocked by synstatin, a novel peptide inhibitor.
Beauvais, DM; Ell, BJ; McWhorter, AR; Rapraeger, AC
The Journal of Experimental Medicine null
Qi Cui et al.
Nature communications, 7, 10637-10637 (2016-02-04)
Glioblastomas have been proposed to be maintained by highly tumorigenic glioblastoma stem cells (GSCs) that are resistant to current therapy. Therefore, targeting GSCs is critical for developing effective therapies for glioblastoma. In this study, we identify the regulatory cascade of
Laura di Blasio et al.
Journal of cell science, 128(5), 863-877 (2015-01-16)
Non-amoeboid cell migration is characterised by dynamic competition among multiple protrusions to establish new adhesion sites at the cell's leading edge. However, the mechanisms that regulate the decision to disassemble or to grow nascent adhesions are not fully understood. Here

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service