8.55053
Dnp NovaTag® resin
Novabiochem®
Synonym(s):
N-Dnp-NÆ-Fmoc-ethylenediamine MPB-AM resin
About This Item
Recommended Products
Quality Level
product line
NovaSyn® TG
Novabiochem®
form
beads
reaction suitability
reaction type: Fmoc solid-phase peptide synthesis
manufacturer/tradename
Novabiochem®
application(s)
peptide synthesis
functional group
amine
storage temp.
15-25°C
General description
Associated Protocols and Technical Articles
Protocols for Loading of Peptide Synthesis Resins
Linkage
Analysis Note
Appearance of substance (visual): beads
Loading (Photometric determination of the Fmoc-chromophore liberated upon treatment with DBU/DMF): 0.30 - 0.60 mmol/g
Swelling Volume (in DMF): lot specific result
The polymer matrix is copoly styrene - 1 % DVB 100 - 200 mesh
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service