Skip to Content
Merck
All Photos(1)

Key Documents

8.51011

Sigma-Aldrich

MSNT

Novabiochem®

Synonym(s):

MSNT, 1-(Mesitylene-2-sulfonyl)-3-nitro-1H-1,2,4-triazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H12N4O4S
CAS Number:
Molecular Weight:
296.30
MDL number:
UNSPSC Code:
12352005

product line

Novabiochem®

Assay

≥95.0% (HPLC)

form

powder

reaction suitability

reaction type: Coupling Reactions

manufacturer/tradename

Novabiochem®

storage temp.

15-25°C

InChI

1S/C11H12N4O4S/c1-7-4-8(2)10(9(3)5-7)20(18,19)14-6-12-11(13-14)15(16)17/h4-6H,1-3H3

InChI key

SFYDWLYPIXHPML-UHFFFAOYSA-N

General description

This versatile reagent was originally used as a coupling reagent in phosphotriester oligonucleotide synthesis [1-3]. In recent years, it has been increasingly used for the esterification of Fmoc-amino acids onto solid phase synthesis supports [4-12]. It has proved particularly effective in difficult cases involving unreactive linkers [8, 9] or enantiomerization prone amino acids [7]. In an investigation [6] into ester formation, in which the alcohol or carboxylic acid component was immobilized on a solid support, MSNT was found to give the best results of all the reagents tested (DCC, DCC/HOBt, DCC/DMAP, Ph3P/DEAD).

The product number for this product was previously 01-62-0021.

To obtain a certificate of analysis (CoA) of a lot that begins with the letter “A”, please select the option in the right hand menu “Request a COA for Lot#s starting with A”.

Linkage

Replaces: 01-62-0021

Analysis Note

Colour (visual): white to slight yellow to beige
Appearance of substance (visual): powder
Identity (IR): passes test
Assay (HPLC, area%): ≥ 95.0 % (a/a)

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service