8.08390
Tributyltin chloride
for synthesis
Synonym(s):
Tributyltin chloride, Chlorotributyltin, Chlorotributylstannane
About This Item
Recommended Products
vapor pressure
<0.01 hPa ( 20 °C)
autoignition temp.
>150 °C
potency
129 mg/kg LD50, oral (Rat)
mp
-9 °C
transition temp
flash point 108 °C
density
1.20 g/cm3 at 20 °C
storage temp.
2-30°C
InChI
1S/3C4H9.Cl.Sn/c3*1-3-4-2;;/h3*1,3-4H2,2H3;;
InChI key
VDWSDASDKIBOHS-UHFFFAOYSA-N
Analysis Note
Density (d 20 °C/ 4 °C): 1.199 - 1.204
Identity (IR): passes test
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2 - Skin Sens. 1B - STOT RE 1
Storage Class Code
6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK
WGK 3
Flash Point(F)
226.4 °F
Flash Point(C)
108 °C
Regulatory Listings
Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.
EU REACH Annex XVII (Restriction List)
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service