Skip to Content
Merck
All Photos(1)

Key Documents

6.17597

Supelco

Cadmium nitrate tetrahydrate

purified grade

Synonym(s):

Cadmium nitrate tetrahydrate, Nitric acid, cadmium salt tetrahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cd(NO3)2 · 4H2O
CAS Number:
Molecular Weight:
308.48
Beilstein:
8156730
MDL number:
UNSPSC Code:
12352302
grade:
purified grade

grade

purified grade

mp

59.5 °C (lit.)

SMILES string

[H]O[H].[H]O[H].[H]O[H].[H]O[H].[O-][N+](=O)O[Cd]O[N+]([O-])=O

InChI

1S/Cd.2NO3.4H2O/c;2*2-1(3)4;;;;/h;;;4*1H2/q+2;2*-1;;;;

InChI key

QOYRNHQSZSCVOW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service