Skip to Content
Merck
All Photos(1)

Documents

235423-M

Millipore

Caspase-3 Inhibitor I, Cell-Permeable

The Caspase-3 Inhibitor I, Cell-Permeable controls the biological activity of Caspase-3. This small molecule/inhibitor is primarily used for Cancer applications.

Synonym(s):

Caspase-3 Inhibitor I, Cell-Permeable, CPP32/Apopain Inhibitor, Cell-permeable, Cell-permeable, DEVD-CHO

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C94H158N20O27
Molecular Weight:
2000.38
UNSPSC Code:
12352200

Quality Level

Assay

≥97% (HPLC)

form

lyophilized solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

solubility

DMSO: 5 mg/mL

storage temp.

−20°C

General description

A cell-permeable inhibitor of caspase-3, as well as caspase-6, caspase-7, caspase-8, and caspase-10. The C-terminal DEVD-CHO sequence of this peptide is a highly specific, potent, and reversible inhibitor of caspase-3 (Ki<1 nM) that has also been shown to strongly inhibit PARP cleavage in cultured human osteosarcoma cell extracts (IC50 = 200 pM). The N-terminal sequence (amino acid residues 1-16) corresponds to the hydrophobic region (h-region) of the signal peptide of Kaposi fibroblast growth factor (K-FGF) and confers cell-permeability to the peptide. A 5 mM (1 mg/100 l) solution of Caspase-3 Inhibitor I, Cell-permeable in DMSO is also available.
A cell-permeable inhibitor of caspase-3. The C-terminal DEVD-CHO sequence of this peptide is a highly specific, potent and reversible inhibitor of caspase-3 (Ki<1 nM) that has also been shown to strongly inhibit PARP cleavage in cultured human osteosarcoma cell extracts (IC50 = 0.2 nM). The N-terminal sequence (residues 1-16) corresponds to the hydrophobic region (h-region) of the signal peptide of Kaposi fibroblast growth factor (K-FGF) and confers cell-permeability to the peptide.

Biochem/physiol Actions

Primary Target
caspase-3
Target IC50: 200 pM for inhibiting PARP cleavage in cultured human osteosarcoma cell extracts

Warning

Toxicity: Standard Handling (A)

Sequence

Ac-Ala-Ala-Val-Ala-Leu-Leu-Pro-Ala-Val-Leu-Leu-Ala-Leu-Leu-Ala-Pro-Asp-Glu-Val-Asp-CHO

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 1 month at -20°C.

Other Notes

Thornberry, N.A., and Lazebnik, Y. 1998. Science 281, 1312.
Lin, Y.-Z., et al. 1996. J. Biol. Chem. 271, 5305.
Nicholson, D.W. 1996. Nature Biotechnol. 14, 297.
Schlegel, J., et al. 1996. J. Biol. Chem.271, 1841.
Lin, Y.-Z., et al. 1995. J. Biol. Chem.270, 14255.
Nicholson, D.W., et al. 1995. Nature 376, 37.
Tewari, M., et al. 1995. Cell 81, 801.
Lazebnik, Y.A., et al. 1994. Nature371, 346.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service