Skip to Content
Merck
All Photos(1)

Documents

1.15326

Supelco

TLC plates, Silica gel 60

pkg of 100 plates, plate L × W 2.5 cm × 7.5 cm, glass support

Synonym(s):

Silica Gel 60 TLC Plates

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115711
NACRES:
NB.21

material

glass support
silica gel 60 matrix

Quality Level

feature

binder Organic Polymer
fluorescent indicator: no

packaging

pkg of 100 plates

technique(s)

thin layer chromatography (TLC): suitable

layer thickness

250 μm

plate L × W

2.5 cm × 7.5 cm

particle size

10-12 μm

pore size

60 Å medium pore diameter

storage temp.

2-30°C

General description

100 Glass plates 2.5 x 7.5 cm
Our classical silica TLC plates are based on a combination of our proven silica gel 60 and the addition of a unique polymeric binder resulting in a very adherent and hard surface that will not crack or blister and even allow writing with a pensil on the surface without risk to damage the layer.

Application


  • Protein-Polymer Conjugates Synthesized Using Water-Soluble Azlactone-Functionalized Polymers: Highlighting the versatility of triethylene glycol monomethyl ether in polymer synthesis, this research develops conjugates for receptor-specific cellular uptake, crucial for targeted drug delivery systems (Kim et al., 2019).

  • Hydrogen-bonded six-component assembly for capsule formation: Utilizing triethylene glycol monomethyl ether, this study creates complex capsule structures with applications in material science, particularly for photoresponsive systems (Togari et al., 2018).

  • An ultra-small thermosensitive nanocomposite with a Mo(154)-core: This research incorporates triethylene glycol monomethyl ether in the development of a comprehensive platform for NIR-triggered photothermal-chemotherapy, marking a significant advancement in cancer treatment technologies (Zhang et al., 2018).

  • Synthesis of Polyester Dendritic Scaffolds for Biomedical Applications: Featuring triethylene glycol monomethyl ether, this article explores its use in the synthesis of polyester dendritic scaffolds, enhancing biomedical applications such as tissue engineering and regenerative medicine (Sadowski et al., 2016).



Linkage

Replaces: 15326-1; 15326

Analysis Note

Specific surface area (according to BET; 5-Pt. measurement): 480 - 540 m²/g
Pore volume (N₂-isotherm): 0.74 - 0.84 ml/g
d 50 (laser diffraction, size distribution): 9.7 - 11.7 µm
Layer thickness: 210 - 270 µm
Deviation of layer thickness per plate: ≤ 35 µm
Chromatographic testing:
colour test
hRf-values
- bleu vif organol, colour test, lipophile:
11 - 25
- Ceres Black G, colour test, lipophile: 34 - 48
- ceres Violet brn, colour test, lipophile: 52 - 67
separation number (colourtest, lipophile): ≥ 9.0
Typical value determined on a conditioned plate
Eluent: toluene (45% rel. humidity)

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service