Skip to Content
Merck
All Photos(2)

Key Documents

07-293-I

Sigma-Aldrich

Anti-PICK1

from rabbit, purified by affinity chromatography

Synonym(s):

PRKCA-binding protein, Protein interacting with C kinase 1, Protein kinase C-alpha-binding protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat, human

species reactivity (predicted by homology)

mouse (based on 100% sequence homology)

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

mouse ... Pick1(18693)

General description

PRKCA-binding protein (UniProt Q62083; also known as Protein interacting with C kinase 1, Protein kinase C-alpha-binding protein) is encoded by the Pick1 (also known as Prkcabp) gene (Gene ID 18693) in murine species. PICK1 is a scaffolding protein expressed in many tissues, where it regulates trafficking of multiple membrane proteins. The PDZ (PSD-95/Dlg/ZO-1/DHR (Dlg homologous region) domain (a.a. 22-105) mediates PICK1 interaction with proteins containing PDZ-binding motifs, such as PKC and acid sensing ion channels (ASICs), while the lipid-binding AH (arfaptin homology)/BAR (Bin/amphiphysin/Rvs) domain (a.a. 144-357) mediates PICK1 homo-dimerization and membrane localization. Pick1 gene-disruption results in downregulated ASICs current and calcium transients during process of ischemia-reperfusion and protects against primary murine cortical neurons against injury caused by OGD-Rep (oxygen and glucose deprivation followed by reperfusion). PICK1 is also shown to act as a negatively regulator against ASIC1 phosphorylation/activation by PKA in rat pulmonary arterial smooth muscle cells, where PICK1 facilitates calcineurin-mediated ASIC1 dephosphorylation.

Specificity

This rabbit polyclonal antibody targets the C-terminal region of mouse and rat PICK1, as well as human PICK1 spliced isoform 1, but not isoform 2, reported by UniProt (Q62083, Q9EP80, Q9NRD5).

Immunogen

Epitope: C-terminus
GST-tagged recombinant mouse PICK1 C-terminal fragment.

Application

Anti-PICK1, Cat. No. 07-293-I, is a highly specific rabbit polyclonal antibody that targets PRKCA-binding protein and has been tested in Immunohistochemistry (Paraffin) and Western Blotting.
Immunohistochemistry Analysis: A 1:1,000 dilution from a representative lot detected PICK1 in human cerebellum, kidney, and pancreas tissue sections.
Research Category
Neuroscience

Quality

Evaluated by Western Blotting in PC12 cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected PICK1 in 10 µg of PC12 rat pheochromocytoma cell lysate.

Target description

~55 kDa observed. 46.60 kDa (human/mouse) and 46.61 kDa (rat) calculated. The larger-than-calculated band size is consistent with that reported in the literature (Xia, J., et al. (1999). Neuron. 22(1):179 187).

Physical form

Affinity purified.
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service