Skip to Content
Merck
All Photos(1)

Key Documents

05-1100-M

Sigma-Aldrich

Anti-Sulfotyrosine Antibody, Clone Sulfo-1C-A2

clone Sulfo-1C-A2, Upstate®, from mouse

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

culture supernatant

antibody product type

primary antibodies

clone

Sulfo-1C-A2, monoclonal

species reactivity

human, mouse, bovine, porcine

species reactivity (predicted by homology)

all

manufacturer/tradename

Upstate®

technique(s)

ELISA: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG2aκ

General description

Sulfotyrosine is a common post-translational modification of secretory, plasma membrane, and lysosomal proteins. The sulfation reaction is catalyzed by specific sulfotransferase enzymes in the trans-Golgi-network. Tyrosine sulfation occurs in all multicellular organisms and up to 1% of the tyrosines in the total protein content of a cell can be sulfated. Sulfotyrosines mediates many receptor-ligand binding interactions, including the interaction of P-selectin glycoprotein ligand-1 with P-selectin, factor VIII binding to von Willebrand factor, and interactions between thrombin and hirudin. Sulfotyrosine is also involved in the entry of several parasites and viruses; for example, tyrosine sulfation of the HIV-1 co-receptor CCR5 is required for viral entry into host cells, and tyrosine sulfation of the Duffy antigen/receptor for chemokines is crucial for erythrocyte invasion by the malaria parasite Plasmodium vivax.

This clone was derived from the antibody described in Kehoe et al., 2006.

Specificity

The antibody is predicted to recognize any sulfotyrosine, regardless of surrounding amino acid sequence.

Application

Detect Sulfotyrosine with Anti-Sulfotyrosine Antibody, Clone Sulfo-1C-A2 (Mouse Monoclonal Antibody), that has been shown to work in WB, IP, ELISA.

Quality

Assayed using sulfatase treated bovine fibrinogen vs. untreated fibrinogen.

Target description

n/a

Physical form

Format: Purified

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Structure and function of broadly reactive antibody PG16 reveal an H3 subdomain that mediates potent neutralization of HIV-1.
Pejchal, Robert, et al.
Proceedings of the National Academy of Sciences of the USA, 107, 11483-11488 (2010)
Yorihiro Nishimura et al.
PLoS pathogens, 6(11), e1001174-e1001174 (2010-11-17)
Enterovirus 71 (EV71) is one of the major causative agents of hand, foot, and mouth disease, a common febrile disease in children; however, EV71 has been also associated with various neurological diseases including fatal cases in large EV71 outbreaks particularly
Isolation of HIV-1-neutralizing mucosal monoclonal antibodies from human colostrum.
Friedman, James, et al.
Testing, 7, e37648-e37648 (2012)
Juan Xu et al.
PloS one, 8(3), e56114-e56114 (2013-03-09)
The latent membrane protein 1 (LMP1), which is encoded by the Epstein-Barr virus (EBV), is an important oncogenic protein that is closely related to carcinogenesis and metastasis of nasopharyngeal carcinoma (NPC), a prevalent cancer in China. We previously reported that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service