Skip to Content
Merck

COM514239098

Combi-Blocks, Inc.

2,5-Difluoro-3-nitrobenzaldehyde

Synonym(s):

2,5-difluoro-3-nitrobenzaldehyde

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C7H3O3N1F2
CAS Number:
Molecular Weight:
187.1
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C7H3F2NO3/c8-5-1-4(3-11)7(9)6(2-5)10(12)13/h1-3H

Inchi Key

WGHZJFNNDXXIGU-UHFFFAOYSA-N

Purity

96%

Storage temp.

Refrigerated

Country of Origin

CN



Still not finding the right product?

Explore all of our products under 2,5-Difluoro-3-nitrobenzaldehyde




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library