Skip to Content
Merck
All Photos(1)

Documents

M-188

Supelco

MT-45 dihydrochloride solution

1.0 mg/mL in methanol (as free base), certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H32N2 · 2 HCl
CAS Number:
Molecular Weight:
421.45
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

estupefaciente (Spain)

concentration

1.0 mg/mL in methanol (as free base)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

[H]Cl.[H]Cl.C1(C(N2CCN(C3CCCCC3)CC2)CC4=CC=CC=C4)=CC=CC=C1

InChI

1S/C24H32N2.2ClH/c1-4-10-21(11-5-1)20-24(22-12-6-2-7-13-22)26-18-16-25(17-19-26)23-14-8-3-9-15-23;;/h1-2,4-7,10-13,23-24H,3,8-9,14-20H2;2*1H

InChI key

PDTBWROWBIVSFO-UHFFFAOYSA-N

General description

MT-45, also known as IC-6, is a piperazone derivative with analgesic activity comparable to morphine. According to a recent report released by the European Monitoring Centre for Drugs and Drug Addiction (EMCDDA), MT-45 has been associated with deaths in the United States and Sweden, since it emerged on the illicit drug market in 2013. This Certified Spiking Solution® is suitable as starting material for calibrators or controls in MT-45 GC/MS or LC/MS methods for urine drug testing, forensic analysis, or clinical toxicology applications.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 3 - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service