870859P
Avanti
C2:0 L-carnitine
Avanti Research™ - A Croda Brand
Synonym(s):
Acetyl L-carnitine
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Assay
>99% (TLC)
form
powder
packaging
pkg of 1x25 mg (870859P-25MG)
manufacturer/tradename
Avanti Research™ - A Croda Brand
lipid type
bioactive lipids
shipped in
dry ice
storage temp.
−20°C
Packaging
5 mL Amber Glass Screw Cap Vial (870859P-25MG)
Legal Information
Avanti Research is a trademark of Avanti Polar Lipids, LLC
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service